Sklarcut™ Operating Scissors, OR Grade, Sklar
Supplier: Sklar
15-3500 15-3505 15-3510 15-3515
89025-608EA 219.6 USD
89025-608 89025-610 89025-612 89025-614
Sklarcut™ Operating Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are used for dissecting, whereas blunt Operating Scissors are used for cutting materials such as sutures, tubing, or gauze.
Sklarcut™ scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.