Merit™ Mayo Dissecting Scissors, Physician Grade, Sklar
Supplier: Sklar
97-312 97-321 97-325 97-308
10184-408EA 16.27 USD
10184-408 10192-398 10193-888 10193-956
Merit™ Mayo Dissecting Scissors, Physician Grade, Sklar
Scissors
Merit™ Mayo Dissecting Scissors are a versatile instrument, that can be used in multiple procedures and multiple settings.
While the curved instrument is used for cutting or dissecting dense or deep tissue, the straight version is used for cutting surface tissue or sutures. Both may be used interchangeably depending on procedure and physician preference.