RS-SC Series Autoclaves, Revolutionary Science
Supplier: REVOLUTIONARY SCIENCE
RS-SC-50 RS-SC-102 RS-SC-200 RS-SC-200P
10024-988EA 1441.55 USD
10024-988 10024-990 10024-992 10024-994
RS-SC Series Autoclaves, Revolutionary Science
Sterilizers Autoclaves (Steam Sterilizers)
The Saniclave 50 an American made autoclave
The Saniclave 102 is American made. This unit has a simple push button operation. It has a thermally protective polymer skin that eliminates burn risk. There is a low water and under and over temperature alarms for safety. The machine has low water and under and over temperature alarms for safety. It has a corrosion resistant, stainless steel chamber. It is easy to clean. The machine is not designed to use wrapped instruments. This is a medical grade sterilizer good for general medical, dental, chiropody, veterinary, laboratory, tattooing, body piercing, fingernail and cosmetic applications.
Our Saniclave 200 is also designed and manufactured in the USA. The machine has a thermally protective polymer skin eliminates burn risk. The machine is a front loading unit, that is designed for wrapped loads. The machine comes with one support tray and one tray for instruments. The use of distilled water is recommended. This is a medical grade sterilizer good for general medical, dental, chiropody, veterinary, laboratory, tattooing, body piercing, fingernail and cosmetic applications.
The Saniclave 200P is also designed and manufactured in the USA. This front loading autoclave has a built in printer. The machine is designed to use wrapped loads. The machine has a thermally protective polymer skin eliminates burn risk. This autoclave comes with one support tray and one tray for the instruments. It is a medical grade sterilizer good for general medical, dental, chiropody, veterinary, laboratory, tattooing, body piercing, health, fingernail and cosmetic applications.
Our Saniclave 50 has simple push button operation. The machine is top loading, and has a removable basket. There is a thermally protective polymer skin eliminates burn risk. The machine has low water and under and over temperature alarms for safety. It has a corrosion resistant, stainless steel chamber. It is easy to clean. This unit is not designed for use with wrapped instruments. Used most often for veterinary and laboratory applications.
Ordering information: Accessories must be purchased separately. Two year warranty included. The Saniclave 50 is Not FDA approved.