Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Supplier: Thermo Scientific Chemicals
038710-AP
038710-K2
038710-K7
038711-K2
038711-K7
038712-AP
038712-K2
038713-AP
038714-K7
040439-K7
040446-K2
040446-K7
042415-K7
042416-K7
042417-AP
042418-AP
042419-AP
042419-K2
042420-K2
042420-K7
042423-K2
042425-AP
042425-K2
038713-K2
042421-AP
042421-K7
042422-K7
038711-AP
038713-K7
042416-AP
042416-K2
042417-K2
042419-K7
042423-AP
042424-AP
042424-K2
040439-K2
040446-AP
040447-K2
042415-AP
042415-K2
042418-K2
042420-AP
042421-K2
042422-AP
042422-K2
042424-K7
038713-M4
038709-AP
038709-K7
038712-K7
038714-AP
038714-K2
040439-AP
040447-AP
040447-K7
042417-K7
042418-K7
042425-K7
038709-K2
046307-AP
046307-K2
046307-K7
AA38710-APEA
46.23
USD
AA38710-AP
AA38710-K2
AA38710-K7
AA38711-K2
AA38711-K7
AA38712-AP
AA38712-K2
AA38713-AP
AA38714-K7
AA40439-K7
AA40446-K2
AA40446-K7
AA42415-K7
AA42416-K7
AA42417-AP
AA42418-AP
AA42419-AP
AA42419-K2
AA42420-K2
AA42420-K7
AA42423-K2
AA42423-K7
AA42425-AP
AA42425-K2
AA38713-K2
AA42421-AP
AA42421-K7
AA42422-K7
AA38711-AP
AA38713-K7
AA42416-AP
AA42416-K2
AA42417-K2
AA42419-K7
AA42423-AP
AA42424-AP
AA42424-K2
AA40439-K2
AA40446-AP
AA40447-K2
AA42415-AP
AA42415-K2
AA42418-K2
AA42420-AP
AA42421-K2
AA42422-AP
AA42422-K2
AA42424-K7
AAA38713-M4
AA38709-AP
AA38709-K7
AA38712-K7
AA38714-AP
AA38714-K2
AA40439-AP
AA40447-AP
AA40447-K7
AA42417-K7
AA42418-K7
AA42425-K7
AA38709-K2
AA46307-AP
AA46307-K2
AA46307-K7
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Buffers
pH Calibration Buffers
NIST Traceable.
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD03458582
Calibration of pH meters
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...