Dimethyl sulfoxide-[D₆] (99.9% D) for NMR spectroscopy
Supplier: CAMBRIDGE ISOTOPE LABS INC MS
Synonyms:
Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6
DLM-10-5
DLM-10-10X0.75
DLM-10-10X0.6
DLM-10-25
DLM-10-50
DLM-10-1000
DLM-10-10X1
DLM-10-10
DLM-10-100
101322-304EA
89
USD
101322-304
101322-312
101322-294
101322-276
101322-296
101322-306
101322-284
101322-278
101322-286
103384-560
Dimethyl sulfoxide-[D₆] (99.9% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula:
D₃CS(O)CD₃ MW: 84.09 g/mol Boiling Pt: 189 Melting Pt: 20.2 Density: 1.19 g/cm³ (20 °C) Flash Pt: 82 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 EINECS: 218-617-0 |
Specification Test Results
Chemical Purity | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...