Latex Examination Gloves, Chemo Rated, Tronex®
Supplier: Tronex
3044STR-10 3044STR-20 3088STR-10 3088STR-20
10153-008BX 70.94 USD
10153-008 10153-010 10153-012 10153-014 10153-016 10153-018 10153-020
Latex Examination Gloves, Chemo Rated, Tronex®
Gloves
Superior latex material formulation with polymer coating provides enhanced softness, elasticity, and reduced protein content, while offering donning ease.
  • Sterilized with gamma radiation at minimum 25 kGy
  • Fully textured for sustained grip
Ordering information: Packaged in individual 'easy-peel' pouches for portability, as well as to resist moisture, reduce tears, and maintain sterile integrity, as either sterile singles (3044STR Series) or pairs (3088STR Series).