HyFlex® 11-735 Medium Duty 10-Gauge Gloves, Palm Dipped, Ansell
Supplier: Ansell Healthcare
11735060 11735090 11735100 11735070 11735080
10002-096CS 2496.68 USD
10002-096 10001-978 10001-980 10001-982 10002-042 10002-044
HyFlex® 11-735 Medium Duty 10-Gauge Gloves, Palm Dipped, Ansell
Gloves
The HyFlex® 11-735 glove features a 10G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN Cut Level 5 (ANSI Level 4) protection for the ultimate in cut resistance.
  • Silicone-free
  • Increased worker confidence when handling small, sharp parts
  • Exceptional abrasion resistance
  • Minimal shrinkage after laundering
The comfortable and flexible plaited liner also features an optimized Lycra® content that is soft and cool on the skin for all-day comfort.
The exceptional precision, dexterity, and flexibility when handling small,More Product Information