Sodium thiosulfate 0.02 N in aqueous solution

Supplier: AQUA SOLUTIONS MS

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

8798-1L 8798-4L 8798-4LC
101223-042EA 63.05 USD
101223-042 101223-854 101225-808
Sodium thiosulfate 0.02 N in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Actual Normality 0.0200 ± 0.0005 Normal (20° C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR