Acetone ≥99.5% ACS
Supplier: AQUA SOLUTIONS MS
|
Danger
|
Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
A1494-500ML
A1494-20L
A1494-1L
A1494-4L
101204-556EA
97.99
USD
101204-556
101204-816
101204-802
101203-932
Acetone ≥99.5% ACS
Acetone
Formula:
CH₃COCH₃ MW: 58.08 g/mol |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 UN: 1090 ADR: 3,II |
Specification Test Results
Assay | 99.5% Min. |
Solubility in water | Passes test |
Aldehyde (as HCHO) | Max. 0.002% |
Isopropyl Alcohol | Max. 0.05% |
Color (APHA) | Max. 10 |
Evaporation residue | Max. 0.001% |
Titrable acid | Max. 0.0003 meq/g |
Titrable base | Max. 0.0006 meq/g |
Substances Reducing Permanganate | Passes test |
Water | Max. 0.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...