1-Adamantanemethylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Adamantan-1-ylmethanamine, 1-Aminomethyladamantane
H66197-03
H66197-06
H66197-MD
AAH66197-03EA
133
USD
AAH66197-03
AAH66197-06
AAH66197-MD
1-Adamantanemethylamine 97%
1-Adamantanemethylamine
1-Adamantanemethylamine, 97%
Purity: 97%
CAS No.: 17768-41-1
Molecular Formula: C11H19N
Formula Weight: 168.28
Flash Point: 92°(198°F)
Physical Form: Liquid
Density: 0.933
Refractive Index: 1.5120
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Boiling Point: 83-85°/0.3mm
UN No.: 2735
MDL No.: MFCD00074750
Formula:
C₁₁H₁₉N MW: 165.28 g/mol Boiling Pt: 83…85 °C (0.3 mmHg) Density: 0.933 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00074750 CAS Number: 17768-41-1 EINECS: 241-752-1 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...