Palladium : Platinum (2.5:2.5)% on carbon, paste Type 122
Supplier: Thermo Scientific Chemicals
|
Warning
|
H36548-06
H36548-22
H36548-14
AAH36548-22EA
934.62
USD
AAH36548-22
AAH36548-14
AAH36548-06
Palladium : Platinum (2.5:2.5)% on carbon, paste Type 122
Palladium : Platinum
Formula:
Pd:Pt Storage Temperature: Ambient |
MDL Number:
MFCD03613605 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...