Platinum 10% on activated carbon paste Type 128M
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Platinium
H36044-06
H36044-22
H36044-14
AAH36044-22EA
1975.01
USD
AAH36044-22
AAH36044-14
AAH36044-06
Platinum 10% on activated carbon paste Type 128M
Platinum
Formula:
Pt MW: 195.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 ADR: 4.2,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...