4-Amino-2-bromobenzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H61247-03 H61247-06
AAH61247-03EA 54.2 USD
AAH61247-03 AAH61247-06
4-Amino-2-bromobenzonitrile 95%
4-Amino-2-bromobenzonitrile

4-Amino-2-bromobenzonitrile, 95%


Purity: 95% CAS No.: 53312-82-6 Molecular Formula: C7H5BrN2 Formula Weight: 197.03 Physical Form: Powder Appearance: Yellow UN No.: 3439 MDL No.: MFCD03695814

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03695814
CAS Number: 53312-82-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR