Cerium(IV) oxide 20% in water, nanoparticle dispersion <5.0nm APS Number Weighted, high pH

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

047240-22 047240-36
AA47240-22EA 206 USD
AA47240-22 AA47240-36
Cerium(IV) oxide 20% in water, nanoparticle dispersion <5.0nm APS Number Weighted, high pH
Cerium (IV) oxide

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH, 5.0nm APS Number Weighted


CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: Liquid Density: 1.23 Appearance: Red-brown Odor: Odorless pH: 8.0 MDL No.: MFCD00010933

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 1.23
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR