MAS™ DOA TOTAL Controls, Thermo Scientific
Supplier: Thermo Scientific
MAS™
DOAT-2 DOAT-4 DOAT-1 DOAT-MP DOAT-6 DOAT-5 DOAT-3
89209-138PK 1652.1 USD
89209-138 89209-136 89209-140 89209-134 89209-144 89209-132 89209-142
MAS™ DOA TOTAL Controls, Thermo Scientific
Controls and Calibration Verification Materials
The MAS™ DOA TOTAL Controls are liquid, ready-to-use urine toxicology controls designed for monitoring immunoassay screening procedures
Liquid, ready-to-use; Human urine based matrix; GC/MS confirmation values available.
Analytes: Benzoylecogonine, Buprenorphine, Cotinine, d-methamphetamine, EDDP, Ethanol, Ethyl Glucuronide, LSD, Mehtadone, Methaqualone, Morphine-free, Nitrazepam, Nortriptyline, Oxazepam, Oxycodone, Phencyclidine, Propoxyphene, Secobarbital, 11-nor-9-COOH-D9-THC.
The new MAS™ DOA TOTAL Control is a multi-constituent urine toxicology control offering 19 analytes with 4 distinct levels at drug concentrations 25% below and above commonly used screening and SAMHSA cutoffs. A drug-free level and high positive level are also available (6 levels total).