Spectroscopy Light Sources
Supplier: United Scientific Supplies
LAMPM01 PFB001-PS220 LAMPS01 LAWPS01
470228-030EA 390.3 USD
470228-030 470218-720 470218-718 470218-716
Spectroscopy Light Sources
Lights Spectrum Lamps
Various type of light sources for spectroscopy experiments.
  • Predicatable spectral lines
  • Includes a closable shield block
Perfect for spectroscopy and other optics experiments. Sodium and mercury light sources have predictable and unique spectral lines which make them perfect for calibrating your spectrometers or other equipment. Each lamp house includes a closable shield to block out light when it is not needed.
Ordering information: The power supply, available separately, can power one lamp at a time.