WHEATON® Headspace Vials, Crimp Top, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225278-01 W225280-01 W225280-02 W225280-03 225278 225277 225280
14227-790CS 288.38 USD
14227-790 14227-794 14227-796 14227-798 14227-800 66021-220 80700-770 80700-772
WHEATON® Headspace Vials, Crimp Top, DWK Life Sciences
Vials
Manufactured from clear type I borosilicate glass, these headspace vials accept 20 mm aluminum seals.
A variety of aluminum seal styles and septa materials are available.
Ordering information: Vials can be purchased separately, or together with seals in convenience packs.