Buck Neurological Hammer
Supplier: AMERICAN 3B SCIENTIFIC SE
1017424
470119-650EA 10.25 USD
470119-650
Buck Neurological Hammer
Mallets and Hammers
A Standard For Sensation Evaluation.
  • Double-Sided Percussion Head
  • Hollow Shaft Contains Point and Brush
The hammer's hollow shaft contains a removable point and brush used for cutaneous sensation evaluations.