Anti-AKT3 Mouse Monoclonal Antibody [clone: 16C8.E4.G7.E9]

Supplier: Rockland Immunochemicals
K-BR8 K-BR8
10799-936EA 179 USD
10799-936 CA10799-936
Anti-AKT3 Mouse Monoclonal Antibody [clone: 16C8.E4.G7.E9]
Antibodies
Combo Pack: This primary and secondary antibody pair comes with matched antibody pairs to detect and quantify endogenous protein levels of human AKT2. AKT2 Antibody detects AKT2 which is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and rac (related to A and C kinases). AKT is a key regulator of many signal transduction pathways. AKT Exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. Anti-AKT2 Antibody is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 16C8.E4.G7.E9
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR