VerityMAX™ DNA Polymerase Master Mix, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5031-0080R 31-5031-0400R
10002-220EA 115.5 USD
10002-220 10002-222
VerityMAX™ DNA Polymerase Master Mix, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VerityMAX DNA Polymerase Master Mix contains a proprietary blend of our VerityPfu and Classic++ Hot Start Taq DNA polymerases, engineered and optimized specifically for generating amplicons of up to 35 kb in length with 2.5x higher fidelity than standard Taq polymerase
Resultant PCR products are both A-tailed and blunt-end finished. This enzyme is suitable for a wide variety of fast or standard PCR protocols when additional speed, sensitivity, specificity and yields are desired.
VerityMAX DNA Polymerase Master Mix accurately and reliably amplify DNA from complex GC/AT rich genomic DNA, plasmid templates and crude samples. It is suitable for multiplexing and colony PCR, especially long-range or difficult PCR.
Hot start technology improves yields by minimizing or eliminating primer dimer formation and non-specific amplification