Ward's® Soil Types Collections
Supplier: Avantor
WARD470025-306
470025-306EA 280.69 USD
470025-306
Ward's® Soil Types Collections
Educational Materials Earth Science Educational Materials Geology Learning Activities
This collection is useful for ecological as well as geological studies.
  • Carefully selected and sourced soils
  • Provides compete representation of soil classes
Twenty specimens of sandy, clayey, loamy, calcareous, and organic soils, plus residual soils, provide a complete representation of soil classes. This set is housed in glass jars, each with its own labeled plastic tray. An identification list with names and localities is also included.