Pierce™ Vacuum Hydrolysis Tubes, Thermo Scientific
Supplier: Thermo Scientific
Pierce
29572 29570 29571
PI29571EA 364.85 USD
PI29571 PI29570 PI29572
Pierce™ Vacuum Hydrolysis Tubes, Thermo Scientific
Tubes Test Tubes
For fast, effective protein and peptide hydrolysis.
  • Fits into Reacti-Block aluminium heating blocks
  • Sufficient for samples up to 0.35 ml
  • Optimal hydrolysis control
The upper temperature limit of the vacuum hydrolysis tubes is 260 °C, however, do not heat the tubes greater than 100 °C in an oven.