Basic T Lymphocyte Backbone Panel [clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14]Basic T Lymphocyte Backbone Panel

Supplier: Tonbo Biosciences
91-0002-U025 91-0002-U100
10002-344EA 315.7 USD
10002-344 10002-346
Basic T Lymphocyte Backbone Panel [clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14]Basic T Lymphocyte Backbone Panel
Antibodies
The Mouse Basic T Lymphocyte backbone panel is a six-color combination of anti-mouse antibodies. This panel can serve as a backbone for identifying various T lymphocyte subsets, including naive and memory T cell populations. This panel's configuration leaves room for GFP or additional drop-in markers, if desired. Violet laser (405 nm) required for detection of CD3e violetFluorTM 450

Contains:
1 x violetFluor™ 450 Anti-Mouse CD3 (17A2)
1 x APC-Cyanine7 Anti-Mouse CD4 (RM4-5)
1 x redFluor™ 710 Anti-Mouse CD8a (53-6.7)
1 x PE-Cyanine7 Anti-Mouse CD25 (PC61.5)
1 x PerCP-Cyanine5.5 Anti-Human/Mouse CD44 (IM7)
1 x APC Anti-Mouse CD62L (L-Selectin) (MEL-14)

Type: Primary
Antigen: CD3, CD4, CD8a; CD25, CD44, CD62L
Clonality: Monoclonal
Clone: 17A2, RM4-5, 53-6.7, PC61.5, IM7, MEL-14
Conjugation: violetFluor® 450, APC (Allophycocyanin)-Cy7, redFluor® 710, PE-Cy7, PerCP-Cy5.5, APC (Allophycocyanin)
Epitope:
Host:
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR