Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]

Supplier: Tonbo Biosciences
20-0108-T025 20-0108-T100 20-0108-T500
10002-102EA 106.7 USD
10002-102 10002-104 10002-106
Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]
Antibodies
The SN5c antibody is specific for human CD10, a 100 kDa type II transmembrane protein also known as Common Acute Lymphoblastic Leukemia Antigen (CALLA). In normal cells, it is expressed on early B and T lymphoid precursors, neutrophils and on various epithelia. CD10 is also expressed on several lymphoma cells including Burkitts, acute lymphoblastic, and follicular germinal center lymphomas. CD10 functions as an endopeptidase and is involved in B cell development, as well as a mediator of neutrophil inflammatory responses.

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: SN5c
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR