Sodium dihydrogen phosphate monohydrate USP endotoxin tested
Supplier: Avantor Performance Materials
Synonyms:
Sodium dihydrogen orthophosphate 1-hydrate
VWRVVWRB0823-12KG
VWRVVWRB0823-1KG
VWRB0823-1KGEA
478.59
USD
VWRB0823-1KG
VWRB0823-12KG
Sodium dihydrogen phosphate monohydrate USP endotoxin tested
Sodium dihydrogen phosphate monohydrate
Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.
Formula:
NaH₂PO₄·H₂O MW: 137.99 g/mol Melting Pt: 100 °C Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 EINECS: 231-449-2 Merck Index: 12,08806 |
Specification Test Results
Aluminum, Calcium & Related Elements | PASS / Fail |
Arsenic | ≤ 8 ppm |
Chloride | ≤ 0.014% |
Endotoxin |
|
Heavy Metals | ≤ 20 ppm |
Identification (A) | PASS / Fail |
Identification (B) | PASS / Fail |
Insoluble Substances | ≤ 0.2 % |
pH (1.0g/20mL Water) @25C | 4.1 - 4.5 |
Purity (Anhydrous Basis) | 98.0 - 103.0% |
Sulfate | ≤ 0.15% |
Water Determination | 10.0 - 15.0% |
Additional information, country of origin | Germany |
Additional information | Material is of non-animal, non-human origin |
Additional information, storage conditions | 15ºC to 30ºC |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...