n-Pentane, anhydrous 99,8, molecular sieves_over
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentane
Argon_Chemseal
047115-AK
047115-K2
AA47115-AKEA
37.94
USD
AA47115-AK
AA47115-K2
n-Pentane, anhydrous 99,8, molecular sieves_over
n-Pentane
- Liquid
Purity: 99.8+%
CAS No.: 109-66-0
Molecular Formula: C5H12
Formula Weight: 72.15
Flash Point: -49°(-57°F)
Physical Form: Liquid
Density: 0.626
Refractive Index: 1.3580
Appearance: Colorless
Odor: Petroleum-like
Melting Point: -130°
Boiling Point: 36°
Beilstein Reference: 969132
UN No.: 1265
MDL No.: MFCD00009498
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36 °C Melting Pt: -130 Density: 0.626 Flash Pt: –49 °C (–57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II Merck Index: 14,7116 |
|
Specification Test Results
Assay Minimum | 99.8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...