Potassium bromide FTIR

Supplier: Thermo Scientific Chemicals
Warning

047111-KD 047111-KM 047111-KN
AA47111-KDEA 263.86 USD
AA47111-KD AA47111-KM AA47111-KN
Potassium bromide FTIR
Potassium bromide

  • Powder in 0.5g Packets


CAS No.: 7758-02-3 Molecular Formula: BrK Formula Weight: 119.01 Physical Form: Powder in 0.5g Packets Density: 2.75 Refractive Index: 1.559 Odor: Odorless Melting Point: 730° Boiling Point: 1435° MDL No.: MFCD00011358

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR