Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 100 mesh Ø 0.11 mm (0.0045 in) Type 316
Supplier: Thermo Scientific Chemicals
Danger
046703-FL 046703-GT 046703-HB
AA46703-FLEA 25.9 USD
AA46703-FL AA46703-GT AA46703-HB
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 100 mesh Ø 0.11 mm (0.0045 in) Type 316
Stainless steel
Wire Cloth
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00148499