Molybdenum, gauze woven from wire 40 mesh Ø 0.13 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

046606-FL 046606-GT
AA46606-FLEA 60.5 USD
AA46606-FL AA46606-GT AA46606-HB
Molybdenum, gauze woven from wire 40 mesh Ø 0.13 mm (0.005 in)
Molybdenum

  • Wire Cloth


CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465

Formula: Mo
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR