Molybdenum, gauze woven from wire 10 mesh Ø 0.38 mm (0.015 mm)

Supplier: Thermo Scientific Chemicals

046494-FL 046494-GT 046494-HB
AA46494-FLEA 38.1 USD
AA46494-FL AA46494-GT AA46494-HB
Molybdenum, gauze woven from wire 10 mesh Ø 0.38 mm (0.015 mm)
Molybdenum

  • Wire Cloth


CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465

Formula: Mo
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR