Cobalt ≥99.8% (metals basis), nanopowder APS 25-30 nm

Supplier: Thermo Scientific Chemicals
Danger

46347-06 046347-06 046347-14 46347-14
AA46347-14EA 360.99 USD
CAAA46347-14 CAAA46347-06 AA46347-14 AA46347-06
Cobalt ≥99.8% (metals basis), nanopowder APS 25-30 nm
Cobalt
Formula: Co
MW: 58.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay Minimum 99.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR