Quartz Tubing, Fused, Thermo Scientific
Supplier: Thermo Scientific Chemicals
042272-NK 042274-NT 042272-NT 042274-NK 042273-NT 042275-NK 042275-NT 042276-NK 042276-NT 043645-NT
AA42272-NKEA 34.5 USD
AA42272-NK AA42274-NT AA42272-NT AA42273-NK AA42274-NK AA42273-NT AA42275-NK AA42275-NT AA42276-NK AA42276-NT AA43645-NK AA43645-NT
Quartz Tubing, Fused, Thermo Scientific
Tubing Transfer Tubing
These are manufactured by fusing naturally occuring crystalline silica.
  • High chemical impurity
  • High corrosion resistance
  • High melting point
  • Extreme hardness
  • Low coefficient of thermal expansion
  • Excellent electrical insulation quality
Optical transmission from ultra-violet to infra-red.