Glassy carbon, plate, Thickness 1 mm (0.04 in) Type2

Supplier: Thermo Scientific Chemicals

038021-GH 038021-FI
AA38021-GHEA 1521 USD
AA38021-GH AA38021-FI
Glassy carbon, plate, Thickness 1 mm (0.04 in) Type2
Glassy carbon

Glassy carbon plates are widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices


  • High purity
  • Insoluble
  • Corrosion resistance
  • Thermal stability
  • Impermeable structure to both gases and liquids


Used in casting, smelting of metal, hard alloy, powder, glass and also in solar energy applications.

Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR