(S,S)-(-)-Hydrobenzoin 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(1S,2S)-1,2-Diphenylethane-1,2-diol
L14082-06
L14082-14
AAL14082-06EA
128
USD
AAL14082-06
AAL14082-14
(S,S)-(-)-Hydrobenzoin 98+%
(S,S)-(-)-Hydrobenzoin
Purity: 98+%
CAS No.: 2325-10-2
Molecular Formula: C14H14O2
Formula Weight: 214.27
Physical Form: Crystals
Appearance: White
Melting Point: 148-150°
Beilstein Reference: 2330888
Merck Reference: 14,4777
Optical Rotation: -94° (c=1 in methanol)
MDL No.: MFCD00064255
Formula:
C₁₄H₁₄O₂ MW: 214.26 g/mol Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064255 CAS Number: 2325-10-2 Merck Index: 14,4777 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...