HEPES-Buffered Saline (2X, pH 7.0), Low Endotoxin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J67505-AE
J67505-AK
AAJ67505-AEEA
77.4
USD
AAJ67505-AE
AAJ67505-AK
HEPES-Buffered Saline (2X, pH 7.0), Low Endotoxin, Thermo Scientific
Buffers
Cell Culture Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...