Sodium Phosphate, 0.2 M Buffer Soln., pH 7.4, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J67424-AE J67424-AK
AAJ67424-AEEA 77.7 USD
AAJ67424-AE AAJ67424-AK
Sodium Phosphate, 0.2 M Buffer Soln., pH 7.4, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers

Liquid


Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00146206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR