U-18666A 95%
Supplier: Thermo Scientific Chemicals
Solid
Purity: 95%
CAS No.: 3039-71-2
Molecular Formula: C25H41NO2.HCl
Formula Weight: 424.06
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to off-white
Melting Point: 195-196.5
Solubility: Soluble in water at 10mg/ml
MDL No.: MFCD00210908
Formula:
C₂₅H₄₂ClNO₂ Melting Pt: 195…196.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210908 CAS Number: 3039-71-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...