Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 20 nm OD50 (Abs. 524 nm)

Supplier: Thermo Scientific Chemicals

J67386-AMI J67386-XF
AAJ67386-AMIEA 339 USD
AAJ67386-AMI AAJ67386-XF
Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 20 nm OD50 (Abs. 524 nm)
Gold

Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red MDL No.: MFCD00003436

Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR