Glycine, 0.2M Buffer Soln., pH 3.0, Low Endotoxin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J67349-AE
J67349-AK
AAJ67349-AEEA
84.77
USD
AAJ67349-AE
AAJ67349-AK
Glycine, 0.2M Buffer Soln., pH 3.0, Low Endotoxin, Thermo Scientific
Buffers
Cell Culture Buffers
Liquid
CAS No.: 56-40-6
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00008131
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...