Ethidium bromide 98% for molecular biology
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide
J67270-14
AAJ67270-14EA
613
USD
AAJ67270-14
Ethidium bromide 98% for molecular biology
Ethidium bromide
Powder
Formula:
C₂₁H₂₀BrN₃ MW: 394.31 g/mol Melting Pt: ca 261 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011724 CAS Number: 1239-45-8 EINECS: 214-984-6 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...