Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 80 nm (Abs. 457 nm)

Supplier: Thermo Scientific Chemicals

J67252-AC J67252-AE
AAJ67252-ACEA 87.2 USD
AAJ67252-AC AAJ67252-AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 80 nm (Abs. 457 nm)
Silver

Liquid


CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Yellow Odor: Odorless MDL No.: MFCD00003397

Formula: Ag
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR