Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 60 nm (Abs. 430 nm)

Supplier: Thermo Scientific Chemicals

J67208-AC J67208-AE
AAJ67208-ACEA 86.8 USD
AAJ67208-AC AAJ67208-AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 60 nm (Abs. 430 nm)
Silver

Liquid


CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow Odor: Odorless MDL No.: MFCD00003397

Formula: Ag
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR