Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 100 nm OD50 (Abs. 572 nm)

Supplier: Thermo Scientific Chemicals

J67153-AMI J67153-XF
AAJ67153-AMIEA 312 USD
AAJ67153-AMI AAJ67153-XF
Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 100 nm OD50 (Abs. 572 nm)
Gold

Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink Odor: Odorless MDL No.: MFCD00003436

Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR