Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 40 nm (Abs. 416 nm)
Supplier: Thermo Scientific Chemicals
J67090-AC
J67090-AE
AAJ67090-ACEA
86.3
USD
AAJ67090-AC
AAJ67090-AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 40 nm (Abs. 416 nm)
Silver
Liquid
CAS No.: 7440-22-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Yellow
Odor: Odorless
MDL No.: MFCD00003397
Formula:
Ag Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...