Gold 95% supplied in 0.1 mM PBS, nanoparticles 5 nm OD1 (Abs. 515-520 nm)
Supplier: Thermo Scientific Chemicals
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Liquid
Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Formula:
Au Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...