Protease Inhibitor Cocktail I, Animal Free, Thermo Scientific

Supplier: Thermo Scientific
J65358-LQ J65358-LQ
AAJ65358-LQEA 79.4 USD
AAJ65358-LQ CAAAJ65358-LQ
Protease Inhibitor Cocktail I, Animal Free, Thermo Scientific
Cell Culture Supplements and Additives
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: White
UN No.:UN3261
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR