Demecolcine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
Purity: 98+%
CAS No.: 477-30-5
Molecular Formula: C21H25NO5
Formula Weight: 371.43
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Light Sensitive
Appearance: Pale yellow
Melting Point: 186°
Solubility: Soluble in 10mg/ml chloroform. Also soluble in ethanol
Beilstein Reference: 2822892
Merck Reference: 14,2887
UN No.: 2811
MDL No.: MFCD00075459
Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
Formula:
C₂₁H₂₅NO₅ Melting Pt: 186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075459 CAS Number: 477-30-5 EINECS: 207-514-6 UN: 2811 ADR: "6.1",III Merck Index: 14,2887 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...