Cyclopiazonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61594-MA
J61594-MF
AAJ61594-MAEA
209.09
USD
AAJ61594-MA
AAJ61594-MF
Cyclopiazonic acid 98%
Cyclopiazonic acid
Inhibitor of the sarco/endoplasmic reticulum calcium pump, calcium-ATPase
Powder
Applications: Inhibitor of the sarco/endoplasmic reticulum calcium pump, calcium-ATPase
Formula:
C₂₀H₂₀N₂O₃ MW: 336.391 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00167445 CAS Number: 18172-33-3 UN: 2811 ADR: 6.1 III |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...