2-Amino-5-chlorophenol 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chloro-2-hydroxyaniline
H64978-14
H64978-22
AAH64978-14EA
82.8
USD
AAH64978-14
AAH64978-22
2-Amino-5-chlorophenol 97+%
2-Amino-5-chlorophenol
Purity: 97+%
CAS No.: 28443-50-7
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Physical Form: Powder
Appearance: Brown
Melting Point: 145-153°
Beilstein Reference: 386421
UN No.: 2811
MDL No.: MFCD02093863
Formula:
C₆H₆ClNO MW: 143.57 g/mol Melting Pt: 145…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093863 CAS Number: 28443-50-7 EINECS: 249-020-3 UN: 2811 ADR: 6.1 III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...