Pyrrolidin-3-ol hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-3-Hydroxypyrrolidine hydrochloride
H64975-06
H64975-14
AAH64975-06EA
48
USD
AAH64975-06
AAH64975-14
AAH64975-22
Pyrrolidin-3-ol hydrochloride 98%
Pyrrolidin-3-ol hydrochloride
Purity: 98%
CAS No.: 86070-82-8
Molecular Formula: C4H9NO.HCl
Formula Weight: 123.58
Physical Form: Solid
Beilstein Reference: 7029051
MDL No.: MFCD03001754
Formula:
C4H10ClNO Storage Temperature: Ambient |
MDL Number:
MFCD03001754 CAS Number: 86070-82-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...